Aluminium oxide 80-200 Mesh for chromatography, activated

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

AL219-500GM AL219-25KG AL219-2.5KG AL219-45KGBL
700000-820EA 267.93 USD
700000-820 700000-816 700000-814 700000-818
Aluminium oxide 80-200 Mesh for chromatography, activated
Aluminium oxide

Alumina, Activated, 80-200 Mesh, Chromatographic Grade is used for a wide range of adsorbent and catalyst applications. Alumina (aluminum oxides) are suitable for column chromatography and adsorptive filtration.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: Al₂O₃
Boiling Pt: 2980 °C (1013 hPa)
Melting Pt: 2045 °C
Density: 3.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355

Order Now

Specification Test Results

Particle size 80-200 Mesh


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR