Sodium hydroxide solution 50% (w/w) ∼12.5 N in aqueous solution, Reagent Grade
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Danger
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
Sodium Hydroxide, 50 Percent (w/w), Solution, Reagent is commonly known as Caustic soda. This inorganic compound is water soluble, as well as in ethanol and methanol. The dissolution of solid Sodium Hydroxide in water creates a highly exothermic reaction. The concentrated solution is suitable for further dilution or as a raw material feedstock in chemical synthetic transformations. As a Reagent grade item, Spectrum Chemical manufactured Sodium Hydroxide met the toughest regulatory standards for quality and purity
Formula:
NaOH Boiling Pt: 119…135 °C (1013 hPa) Melting Pt: 9…10 °C Density: 1.515 Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...