Sodium hydroxide, soluble suitable for use as excipient, purified

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

S1305-12KG S1305-2.5KG
700000-742EA 535.81 USD
700000-742 700000-744
Sodium hydroxide, soluble suitable for use as excipient, purified
Sodium hydroxide

Sodium Hydroxide, Flakes, Purified is also known as caustic soda is used as a strong chemical base in the manufacture of paper, pulp, textiles, soaps, and detergents, as well as a drain cleaner. The Purified Grade describes chemicals of good quality where there are no official standards and is normally limited to inorganic chemicals.

Formula: NaOH
Boiling Pt: 1390 °C (1013 hPa)
Melting Pt: 323 °C
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Merck Index: 14,08627

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR