Sodium hydroxide, coarse granules, Technical Grade suitable for use as excipient
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Danger
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
S1302-12KG
S1302-2.5KG
S1302-500GM
700000-736EA
564.18
USD
700000-736
700000-738
700000-740
Sodium hydroxide, coarse granules, Technical Grade suitable for use as excipient
Sodium hydroxide
Sodium Hydroxide, Coarse Granular, Technical is a strong base that is also used in manufacturing of sodium salts and detergents, pH regulation, and organic synthesis. Technical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use. These materials may or may not have a Certificate of Analysis available
Formula:
NaOH Boiling Pt: 1390 °C (1013 hPa) Melting Pt: 323 °C Density: 2.13 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1823 ADR: 8,II REACH: 01-2119457892-27 Merck Index: 14,08627 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...