Sodium hydroxide, coarse granules, Technical Grade suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

S1302-12KG S1302-2.5KG S1302-500GM
700000-736EA 564.18 USD
700000-736 700000-738 700000-740
Sodium hydroxide, coarse granules, Technical Grade suitable for use as excipient
Sodium hydroxide

Sodium Hydroxide, Coarse Granular, Technical is a strong base that is also used in manufacturing of sodium salts and detergents, pH regulation, and organic synthesis. Technical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use. These materials may or may not have a Certificate of Analysis available

Formula: NaOH
Boiling Pt: 1390 °C (1013 hPa)
Melting Pt: 323 °C
Density: 2.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Merck Index: 14,08627

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR