Sulfuric acid 95.0-98.0% NF

Supplier: SPECTRUM CHEMICAL MFG CORP

S1731-2.5LTP S1731-200LTB S1731-2.5LTG S1731-500MLG S1731-2.5LGL-C S1731-2.5LPV-C S1731-20LT S1731-500ML-CS
700000-588EA 366.95 USD
700000-588 700000-590 700000-586 700000-592 75812-370 75812-372 75812-374 75812-376
Sulfuric acid 95.0-98.0% NF
Sulfuric acid

Sulfuric Acid, NF is used as an acidifying agent. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.840
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 1830
ADR: 8 II

Order Now

Specification Test Results

Assay (H₂SO₄) 95.0 - 98.0 %
Residue On Ignition max. 0.005 %
Chloride (Cl) max. 50 ppm
Arsenic (As) max. 1 ppm
Heavy Metals max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR