Paraffin wax, medium flakes

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Wax Paraffin, Paraffin wax

W1061-1LB
700000-500EA 80.9 USD
700000-500
Paraffin wax, medium flakes
Paraffin

Wax Paraffin, Pastilles consists of a mix of hydrocarbon molecules and is derived from coal, petroleum, or oil shale. It can be used in electrical insulation, lubrication and candles.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
MDL Number: MFCD00132833
CAS Number: 8002-74-2
EINECS: 232-315-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR