Paraffin wax, white, cake

Supplier: Spectrum Chemical Mfg. Corp.

W1025-1LB
700000-496EA 93.51 USD
700000-496
Paraffin wax, white, cake
Paraffin

Wax Paraffin, White, Cake consists of a mix of hydrocarbon molecules and is derivable from coal, petroleum, or oil shale. It is used in electrical insulation, lubrication and candles.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Density: 0.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00132833
CAS Number: 8002-74-2
EINECS: 232-315-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR