Paraffin wax, beads

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Wax Paraffin, Paraffin wax

W1018-1LB
700000-494EA 104 USD
700000-494
Paraffin wax, beads
Paraffin

Wax Paraffin, Beads consists of a mix of hydrocarbon molecules and is derivable from coal, petroleum, or oil shale. It is used in electrical insulation, lubrication and candles.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Density: 0.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00132833
CAS Number: 8002-74-2
EINECS: 232-315-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR