Mineral oil heavy USP

Supplier: Spectrum Chemical Mfg. Corp.
Warning

MI500-20LTPL MI500-4LTGL MI500-4CS4 MI500-1LT MI500-500MLG MI500-500MLG MI500-200LTB MI500-200LTB
89078-896EA 1181.92 USD
89078-896 95034-618 95034-616 95034-614 700000-466 95034-620 700000-464 89078-898
Mineral oil heavy USP
Mineral oil

Mineral Oil, Heavy, USP is a lubricant excipient material. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Boiling Pt: 300 °C (1013 hPa)
Density: 0.872 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00131611
CAS Number: 8012-95-1
EINECS: 232-455-8

Order Now

Specification Test Results

SPECIFIC GRAVITY @ 25oC 0.845-0.905
VISCOSITY @40.0 C 34.5-150.0 mm²


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR