Calcium phosphate tribasic, powder NF

Supplier: Spectrum Chemical Mfg. Corp.

CA165-100LBB CA165-500GM CA165-12KG CA165-2.5KG
700000-380EA 5235.35 USD
700000-380 700000-386 700000-382 700000-384
Calcium phosphate tribasic, powder NF
Calcium phosphate tribasic

TRIBASIC CALCIUM PHOSPHATE, POWDER, NF is also referred to as tricalcium phosphate and is a calcium salt of phosphoric acid. It is one of the main byproducts of bone combustion but can also be obtained from inorganic sources such as mineral rock. It is used in baby powder as well as in powdered spices as an anticaking agent. It is also used in gene transfection and in diet supplements for repairing bony defects. Historically tribasic calcium phosphate been used in the production of porcelain. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: Ca₅HO₁₃P₃
Storage Temperature: Ambient
MDL Number: MFCD00010904
CAS Number: 12167-74-7

Order Now

Specification Test Results

Assay (Ca) 34.0 - 40.0 %
Loss On Ignition max. 8.0 %
Water-Soluble Substances max. 0.5 %
Acid-Insoluble Substances max. 0.2 %
Carbonate (CO₃) To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR