Sodium chloride ≥99%, crystals ACS

Supplier: Spectrum Chemical Mfg. Corp.

S1240-500GM S1240-12KG S1240-1KG S1240-2.5KGG
700000-324EA 108.22 USD
700000-324 700000-314 700000-318 700000-320
Sodium chloride ≥99%, crystals ACS
Sodium chloride

Sodium Chloride, Crystal, Reagent, ACS, or table salt, is a white crystalline solid. In cosmetics and personal care products, Sodium Chloride is used in the formulation of oral hygiene products, shampoos, fragrance, skin, hair, nail, cleansing, suntan, makeup and bath products. This grade is specifically used in thickener applications. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness

Formula: NaCl
Boiling Pt: 1413 °C (1013 hPa)
Melting Pt: 801 °C
Density: 2.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR