Lactose, anhydrous NF suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Milk sugar, D-Lactose, Anhydrous Lactose, (2R,3R,4R,5R)-2,3,5,6-tetrahydroxy-4-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal

LA103-2.5KG LA103-500GM LA103-12KG LA103-45KGBL
95033-796EA 570.48 USD
95033-796 95033-798 700000-242 700000-244
Lactose, anhydrous NF suitable for use as excipient
Lactose

Anhydrous Lactose, NF is used as a tablet diluent in the pharmaceutical industry. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: C₁₂H₂₂O₁₁
Boiling Pt: 592 °C (1013 hPa)
Melting Pt: 222.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00151251
CAS Number: 63-42-3
EINECS: 200-559-2
Merck Index: 12,05356

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR