Soybean oil, epoxidised, epoxidised

Supplier: SPECTRUM CHEMICAL MFG CORP

S1179-18KGBL S1179-200LTB S1179-500GM S1179-2.5KG
700000-202EA 1675.6 USD
700000-202 700000-206 700000-208 700000-204
Soybean oil, epoxidised, epoxidised
Soybean oil, epoxidised

Soybean Oil, Epoxidized is used as a plasticizer and stabilizer in polyvinyl chloride (PVC) plastics.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₅₇H₉₈O₁₂
Density: 0.997
Storage Temperature: Ambient
MDL Number: MFCD00163560
CAS Number: 8013-07-8

Order Now

Specification Test Results

Specific gravity at 25 °C 0.988 - 0.998
Acid value 0.50
Color (Alpha) 125
Iodine value 2.00
Moisture 0.10%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR