Cod liver oil USP
Supplier: Spectrum Chemical Mfg. Corp.
Cod Liver Oil, USP is an oil from the liver of Cod fish.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Storage Temperature:
Ambient |
MDL Number:
MFCD00146493 CAS Number: 8001-69-2 |
Specification Test Results
Vitamin A Content | 180 - 750 Ug/G |
Vitamin D Content | 1.5 - 6.25 Ug/G |
Color | to Pass Test |
Specific Gravity @ 25°C | 0.918-0.927 |
Acid Value | To Pass Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...