Inulin

Supplier: Spectrum Chemical Mfg. Corp.

IN145-25GM IN145-100GM IN145-1KG IN145-5GM
700000-022EA 151.3 USD
700000-022 700000-018 700000-020 700000-024
Inulin
Inulin

Inulin, part of a group of natural polysaccharides that belong to a class of dietary fibers known as fructans. Inulin is increasingly used in processed foods as a sugar substitute.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
MDL Number: MFCD00131407
CAS Number: 9005-80-5

Order Now

Specification Test Results

Loss On Drying max. 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR