VWR® JustCast Casting Chamber, Adjustable
Supplier: VWR International
80-GAL-CST
700-0849EA 306.7 USD
700-0849
VWR® JustCast Casting Chamber, Adjustable
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
The JustCast system is a flexible and quick system for casting multiple PerfectBlue™ compatible 'mini' gels at the same time, without the need to tape. Adjustable to fit all gel trays featured in the PerfectBlue™ mini gel series, this system comes fitted with an easy to read bubble level and adjustable feet to ensure a level casting of the gel. At any one time, the system can fit three Mini S, two Mini M or two Mini L trays.