Magid® WeldPro® T2701S Shoulder Split Cow Leather Welding Gloves, Magid
Supplier: Magid Glove
T2701S
69100-598CS 914.13 USD
69100-598
Magid® WeldPro® T2701S Shoulder Split Cow Leather Welding Gloves, Magid
Gloves
Magid WeldPro T2701S Welding Gloves are made of full premium shoulder split cowskin leather for maximum protection during the toughest jobs.
  • Full premium split cowskin leather
  • Jersey/foam lining for max protection
  • Easy-fitting gunn cut pattern
  • Kevlar stitching for added durability
  • Fully welted at all critical seams
Long-lasting welding protection at an affordable price.