VWR® Sterilized Softside Safety Goggles
Supplier: VWR International
69100-210
69100-210CS 1088.04 USD
69100-210
VWR® Sterilized Softside Safety Goggles
Goggles
Softside indirect Cap Safety Goggles feature a green tinted frame with an indirect cap-vented frame for exceptional eye protection and splash resistance and a fog-free lens.
  • Green tinted frame with fog-free lens
  • Indirect cap-vented frame provides splash resistance
  • Soft, clear plastic construction for a comfortable fit and good peripheral vision
These economically priced safety goggles feature a soft construction that delivers a comfortable fit and allows for good peripheral vision. Indirect vented glasses are sterilized in the United States by SterisMore Product Information