Gateway® Safety StarLite Safety Glasses, Magid Glove
Supplier: Magid Glove
4680
69100-192EA 10.2 USD
69100-192
Gateway® Safety StarLite Safety Glasses, Magid Glove
Safety Glasses
Gateway StarLite Glasses are incredibly light with deep, universal-fit temples for side protection equal to that of eyewear with large sideshields. Lenses are scratch resistant, hard coated, 100% polycarbonate and feature revolutionary, dual, 9.75 base curvature for viewing areas with wide, unobstructed vision while blocking more than 99.9% of UV-A and UV-B light.
  • All-around impact protection
  • Popular sunglasses design with full brow protection
  • Deep, universal-fit temples provide side protection equal to that of large sideshields
Lenses also meet or exceed ANSI Z87.1+ standard. Popular design resembles sunglasses helping to ensure worker compliance.