NovaHol™ Cleanroom Detergent, Micronova
Supplier: Micronova
NH1-G
68400-682CS 595.54 USD
68400-682 68400-684
NovaHol™ Cleanroom Detergent, Micronova
Cleaning Agents Detergents and Surface Cleansers
This ready to use, fast drying, low-ionic detergent with added isopropyl alcohol is ideal for use on glass, acrylic, and stainless steel cleanroom surfaces.
  • Ready to use, cleanroom surfactant
  • Fast drying formula with added isopropyl alcohol
  • High evaporation rate
  • Ideal for use on glass, acrylic, stainless surfaces and epoxy
  • Filtered to 0.1 µm; free of mobile ions
  • Low Ionic; low in sodium and potassium
  • Low non-volatile residue levels
  • Biodegradable and free-rinsing
The Micronova NovaHol™ Cleanroom Cleaner is designed with a high evaporation rate, allowing direct application to a variety of surfaces including glass, acrylic, vinyl curtains, softwalls, and stainless steel. The low concentration of isopropyl alcohol in the formula allows it to be used on vinyl and plastic surfaces. Available in an easy to use, 1 quart spray bottle and a 1 gallon bottle.