SATWipes® Sigma™ Wipes, Contec®
Supplier: Contec
SAT-C1-7030/18 SWC30029 SAT-C1-7030 SAT-C1-1090 SAT-C1-100
94025-802CS 506.69 USD
94025-802 33502-126 47734-200 68310-580 68310-574 68310-576
SATWipes® Sigma™ Wipes, Contec®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
The SATWIPES® Sigma™ consists of a perforated roll of saturated wipes which are dispensed through a center-pull canister. The wipes are made of 55% cellulose and 45% polyester creating highly sorbent and durable wipe.
  • Hydraulically entangled fabric creates a soft yet durable wipe
  • Clean wipe offers consistent solvent application
  • Low levels of particles and extractable contaminants to reduce contamination possibility
The isopropanol solvent blends provide safe cleaning solutions in various concentrations. Packaged with refill packs and a reusable pop-upMore Product Information