CONSTIX®, Sealed Foam Swabs, SF-12 Series, Contec®
Supplier: Contec
SF-12
68310-252CS 88.77 USD
68310-252
CONSTIX®, Sealed Foam Swabs, SF-12 Series, Contec®
Swabs Swabs for Cleaning
CONSTIX® SF-12 sealed foam swabs consist of polyurethane foam bonded to a polypropylene handle without the use of adhesives or binders.
  • Small head size enables good access to small features and difficult to reach surfaces
The polyurethane open-cell foam head provides good particle entrapment and removal. The absorbent foam enables solvents, such as isopropanol and ethanol, to be readily absorbed for applications involving solvent delivery or removal. The SF-12 swab is excellent for applying resins and adhesives. Though compatible with many common solvents, the SF-12 swab is not recommended for use with ketones such as acetone or MEK.
Ordering information: Laundered and double bagged.