CONSTIX®, Sealed Foam Swabs, SF-7 Series, Contec®
Supplier: Contec
SF-7
68310-240CS 267.96 USD
68310-240
CONSTIX®, Sealed Foam Swabs, SF-7 Series, Contec®
Swabs Swabs for Cleaning
CONSTIX® SF-7 swabs consist of 100ppi polyurethane foam adhesive-bonded to a flexible nylon tip. The tip is mechanically secured in a tubular polypropylene handle.
  • Flexible head size enables access to small areas and crevices
The polyurethane foam head provides good particles entrapment and removal. The absorbent foam enables solvents, such as isopropanol and ethanol, to be readily sorbed for applications involving solvent delivery and removal.
Though compatible with many common solvents, the SF-7 swab is not recommended for use with ketones such as acetone or MEK.
Ordering information: Laundered and double bagged.