CONSTIX®, Sealed Foam Swabs, SF-5 Series, Contec®
Supplier: Contec
SF-5
68310-236CS 140.46 USD
68310-236
CONSTIX®, Sealed Foam Swabs, SF-5 Series, Contec®
Swabs Swabs for Cleaning
CONSTIX® SF-5 swabs consist of a polyurethane foam head bonded to a flex-tip polypropylene handle without the use of adhesives or binders.
  • Flexible swab head tip provides good surface contact and controlled pressure to surfaces
The polyurethane foam head provides good particle entrapment and removal. The absorbent foam enables solvents, such as isopropanol and ethanol, to be readily sorbed for applications involving solvent delivery or removal. The SF-5 swab is excellent for applying resins and adhesives.
Though compatible with many common solvents, the SF-5 is not recommended for use with ketones such as acetone or MEK.
Ordering information: Laundered and double bagged.