CONSTIX®, Sealed Foam Swabs, SF-3 Series, Contec®
Supplier: Contec
SF-3
68310-232CS 138.97 USD
68310-232
CONSTIX®, Sealed Foam Swabs, SF-3 Series, Contec®
Swabs Swabs for Cleaning
CONSTIX® SF-3 swabs consist of 100ppi sealed open-cell foam head with a polypropylene handle. The swab head is thermally sealed and bonded to the handle without adhesives or binders.
  • Large head is ideal for cleaning and applying coatings
The polyurethane foam head provides good particle entrapment and removal. The SF-3 swab is compatible with many solvents, such as isopropanol and ethanol, but is not recommended for use with ketones, such as acetone or MEK.
Ordering information: Laundered and double bagged.