Polywipe-C Wipers, Contec®
Supplier: Contec
PC-99 PC-99B PC-1212
68310-160CS 863.31 USD
68310-160 68310-162 68310-164
Polywipe-C Wipers, Contec®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Polywipe-C Wipes are made from a standardweight 100% knitted polyester that is knife cut to create a soft edge.
  • Good sorbency makes it compatible with many solvents
  • Abrasion and chemical resistant
Soft edge is ideal for use on surfaces which are vulnerable to scratching. The wipe is exceptionally low in particles and extractable residue making it ideal for critical cleaning. The wipes are laundered and packed in an ISO Class 4 cleanroom.
Certifications: Recommended for ISO Class 5–6 environments.
Ordering information: Wipers 80080-906 and 80080-454 are packaged 150 wipers per bag.