Sodium chloride solution 0.9% for irrigation

Supplier: B BRAUN MEDICAL

R5201-01
68200-070CS 136.66 USD
68200-070
Sodium chloride solution 0.9% for irrigation
Sodium chloride

  • Ideal for sterile overhead irrigation
  • Designed for easy non-drip pouring
  • Not manufactured with PVC or DEHP
  • Lightweight, durable and shatterproof


Ready for immediate use off the shelf, these high-quality solution come in semi-rigid Plastic Irrigation Containers (PIC™).


NDc number: 0264-2201-10

Formula: NaCl
MDL Number: MFCD00003477
CAS Number: 7647-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR