Ringer lactate solution for irrigation, sterile
Supplier: BAXTER IV SOLUTIONS
2F7154
68100-042CS
398.7
USD
68100-042
Ringer lactate solution for irrigation, sterile
Ringer lactate solution
Ringer's sterile solutions for irrigation.
- NDC number: 0338-0114-04
- mEq/L: Na 130, K 4, Ca 2.7, Cl 109, Lactate 28
- mOsmol/L (calc): 273
- pH: 6.5 (6.0–7.5)
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)