Sodium chloride 0,9% USP for irrigation

Supplier: BAXTER IV SOLUTIONS

Synonyms: Saline, NaCl, Sea salt, Halite

2F7123
68100-026CS 375.35 USD
68100-026
Sodium chloride 0,9% USP for irrigation
Sodium chloride

Solutions for irrigation (for Preparation of Slushed Solution)


  • mEq/L: Na 154, Cl 154
  • mOsmol/L (calc): 308
  • NDC Number: 0338-0048-03
  • pH: 5.5 (4.5–7.0)

Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Merck Index: 14,08599

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR