Water USP for irrigation, sterile

Supplier: BAXTER IV SOLUTIONS

Synonyms: Dihydrogen oxide

2F7114
68100-000CS 260.51 USD
68100-000
Water USP for irrigation, sterile
Water

Solutions for irrigation.


  • NDC Number: 0338-0004-04
  • pH 5.5 (5.0–7.0)

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR