Water for injection (WFI)
Supplier: Fresenius Kabi
Synonyms:
Dihydrogen oxide
918510
918520
68099-178CS
11652.06
USD
68099-178
68099-176
68099-180
68099-186
Water for injection (WFI)
Water
Sterile injections comply with USP standards and are bar coded for maximum traceability.
- Preservative-free
Formula:
H₂O |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...