Sodium chloride 0,9% USP

Supplier: Fresenius Kabi

Synonyms: Saline, NaCl, Sea salt, Halite

924830
68099-166PK 114.4 USD
68099-166
Sodium chloride 0,9% USP
Sodium chloride

Sterile injections comply with USP standards and are bar coded for maximum traceability.


  • Preservative-free
  • NDC number: 924-30

Formula: NaCl
MDL Number: MFCD00003477
CAS Number: 7647-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR