Sodium chloride 0,9% USP

Supplier: Fresenius Kabi

Synonyms: Saline, NaCl, Sea salt, Halite

918602 186100 918610 918620
68099-156PK 62 USD
68099-156 68099-162 47729-565 68099-103
Sodium chloride 0,9% USP
Sodium chloride

Sterile injections comply with USP standards and are bar coded for maximum traceability.


  • Preservative-free

Formula: NaCl
MDL Number: MFCD00003477
CAS Number: 7647-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR