Lidocaine hydrochloride 20 mg/ ml USP for injection, sterile

Supplier: Fresenius Kabi

20805 20202
68099-144PK 201.82 USD
68099-144 68099-142
Lidocaine hydrochloride 20 mg/ ml USP for injection, sterile
Lidocaine
Formula: C₁₄H₂₂N₂O
MDL Number: MFCD00026733
CAS Number: 137-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR