Empty Vials for I.V. Injection, Fresenius Kabi
Supplier: Fresenius Kabi
SV30
68099-122PK 101.87 USD
68099-122
Empty Vials for I.V. Injection, Fresenius Kabi
Vials
This vial contains one drop of pyrogen-free water for injection to render the vial sterile upon autoclaving.
Container closure is not made from natural rubber latex.