Chloramphenicol sodium succinate USP for injection, sterile

Supplier: Fresenius Kabi

1115
68099-108PK 932.41 USD
68099-108
Chloramphenicol sodium succinate USP for injection, sterile
Chloramphenicol sodium succinate

Sterile injections comply with USP standards and are bar coded for maximum traceability.


  • Preservative-free

Formula: C₁₅H₁₅Cl₂N₂NaO₈
MDL Number: MFCD00083598
CAS Number: 982-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR