Dextrose 5% in water for injection, sterile
Supplier: BAXTER IV SOLUTIONS
Synonyms:
Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose
2B0089
68001-060CS
1519.82
USD
68001-060
Dextrose 5% in water for injection, sterile
D(+)-Glucose
Sterile solutions for injection.
- Intravenous use
- Subcutaneous use
- mOsmol/L (calc): 252
- NDC Number: 0338-0017-38
- pH: 4.0 (3.2–6.5)
Packaging: Multi Pack contains 16 bags per overwrap pouch. Quad Pack contains four bags per overwrap pouch. Single Pack contains one bag per overwrap pouch.
Formula:
C₆H₁₂O₆ MW: 180.16 g/mol Melting Pt: 146 |
MDL Number:
MFCD00148912 CAS Number: 50-99-7 Merck Index: 14,04459 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...