Mannitol solution 20% in water, Osmitrol® USP for injection

Supplier: BAXTER IV SOLUTIONS

Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol

2D5633Q
68000-986CS 6487.28 USD
68000-986
Mannitol solution 20% in water, Osmitrol® USP for injection
D(-)-Mannitol

Solution for injection


  • Intravenous use
  • Subcutaneous use


mOsmol/L (calc): 1098
NDC Number: 0338-0357-03
pH: 5.0 (4.5–7.0)


Packaging: Multi Pack contains 16 bags per overwrap pouch. Quad Pack contains four bags per overwrap pouch. Single Pack contains one bag per overwrap pouch.

Formula: HOH₂C(CH(OH))₄CH₂OH
MW: 182.17 g/mol
MDL Number: MFCD00064287
CAS Number: 69-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR