Mannitol solution 5% in water, Osmitrol® USP for injection

Supplier: BAXTER IV SOLUTIONS

Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol

2D5604X
68000-976CS 3137.35 USD
68000-976
Mannitol solution 5% in water, Osmitrol® USP for injection
D(-)-Mannitol

Solution for injection


  • Intravenous use
  • Subcutaneous use


NDC number: 0338-0351-04


Packaging: Multi Pack contains 16 bags per overwrap pouch. Quad Pack contains four bags per overwrap pouch. Single Pack contains one bag per overwrap pouch.

Formula: HOH₂C(CH(OH))₄CH₂OH
MW: 182.17 g/mol
MDL Number: MFCD00064287
CAS Number: 69-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR