Ringer lactate solution USP for injection, sterile
Supplier: BAXTER IV SOLUTIONS
2B2322Q
68000-416CS 1212.58 USD
68000-416
Ringer lactate solution USP for injection, sterile
Ringer lactate solution
Ringer's sterile solutions for injection.
  • Intravenous use
  • Subcutaneous use
  • mEq/L: Na 130, K 4, Ca 2.7, Cl 109, Lactate 28
  • mOsmol/L (calc): 273
  • NDC Number: 0338-0117-02
  • pH: 6.5 (6.0–7.5)
For the 5000 mL size of Lactated Ringer's, dosage should be as directed by a veterinarian for intravenous or subcutaneous use or as an irrigating solution.
Packaging: Multi Pack contains 16 bags per overwrap pouch. Quad Pack contains four bags per overwrap pouch. Single Pack contains one bag per overwrap pouch.