Ringer lactate solution in water (+ Dextrose 5%) USP for injection, sterile
Supplier: BAXTER IV SOLUTIONS
2B2073Q
68000-394CS
1042.02
USD
68000-394
Ringer lactate solution in water (+ Dextrose 5%) USP for injection, sterile
Ringer lactate solution
Sterile solutions for injection.
- Intravenous use
- Subcutaneous use
- mEq/L: Na 130, K 4, Ca 2.7, Cl 109, Lactate 28
- mOsmol/L (calc): 525
- NDC Number: 0338-0125-03
- pH: 5.0 (4.0–6.5)
Packaging: Multi Pack contains 16 bags per overwrap pouch. Quad Pack contains four bags per overwrap pouch. Single Pack contains one bag per overwrap pouch.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)