Sodium chloride 3% USP for injection

Supplier: BAXTER IV SOLUTIONS

Synonyms: Saline, NaCl, Sea salt, Halite

2B1353Q
68000-350CS 724.62 USD
68000-350
Sodium chloride 3% USP for injection
Sodium chloride

Sterile solutions for injection.


  • Intravenous use
  • Subcutaneous use
  • mEq/L: Na 513, Cl 513
  • mOsmol/L (calc): 1027
  • NDC Number: 0338-0054-03
  • pH: 5.5 (5.0–7.5)


Packaging: Multi Pack contains 16 bags per overwrap pouch. Quad Pack contains four bags per overwrap pouch. Single Pack contains one bag per overwrap pouch.

Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Merck Index: 14,08599

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR