Dextrose 5% with 0.45% NaCl USP for injection

Supplier: BAXTER IV SOLUTIONS

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

2B1073Q
68000-286CS 550.02 USD
68000-286
Dextrose 5% with 0.45% NaCl USP for injection
D(+)-Glucose
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 146
MDL Number: MFCD00148912
CAS Number: 50-99-7
Merck Index: 14,04459

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR