Water USP

Supplier: BAXTER IV SOLUTIONS

Synonyms: Dihydrogen oxide

2B0306
68000-196CS 250.9 USD
68000-196
Water USP
Water

Sterile Water for injection.


  • Intravenous use
  • Subcutaneous use


NDC Number: 0338-0013-06
pH: 5.5 (5.0–7.0)


Packaging: Multi Pack contains 16 bags per overwrap pouch. Quad Pack contains four bags per overwrap pouch. Single Pack contains one bag per overwrap pouch.

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR