Dextrose solution 10% USP for injection

Supplier: BAXTER IV SOLUTIONS

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

2B0163Q
68000-167CS 883.11 USD
68000-167
Dextrose solution 10% USP for injection
D(+)-Glucose

  • Total calories: 340 kcal/l
  • Specific gravity: 1.03
  • Fill range volume (ml): 1030 to 1070
  • Store at room temperature (25 °C)
  • mOsmol/L (calc): 505
  • NDC Number: 0338-0023-03
  • pH 4


Certifications: USP specified.

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 146
MDL Number: MFCD00148912
CAS Number: 50-99-7
Merck Index: 14,04459

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR