Dextrose 5% in water for injection, sterile

Supplier: BAXTER IV SOLUTIONS

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose

2B0088
68000-152CS 3230.16 USD
68000-152
Dextrose 5% in water for injection, sterile
D(+)-Glucose

Sterile solutions for injection.


  • Intravenous use
  • Subcutaneous use
  • mOsmol/L (calc): 252
  • NDC Number: 0338-0017-31
  • pH: 4.0 (3.2–6.5)


Packaging: Multi Pack contains 16 bags per overwrap pouch. Quad Pack contains four bags per overwrap pouch. Single Pack contains one bag per overwrap pouch.

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 146
MDL Number: MFCD00148912
CAS Number: 50-99-7
Merck Index: 14,04459

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR