ART® XLG Self-Sealing Barrier Pipette Tips, Molecular BioProducts
Supplier: Thermo Scientific
2160G
66666-000CS 1132.73 USD
66666-000
ART® XLG Self-Sealing Barrier Pipette Tips, Molecular BioProducts
Pipette Tips
Tips ensure 100% protection against aerosol and liquid contamination.
  • Pre-sterilized tips
Tips feature an extended length and large orifice for handling fragile cells, viscous samples, and genomic DNA. The ART® barrier traps contaminants, keeping them out of pipettors and samples. This also protects the sensitive inner components of the pipettor, prolonging pipettor life. ART® tips are ideal for nucleic amplification procedures, pipetting of radioactive samples, or any application where critical sample handling is required. Tips are compatible with Eppendorf®, Socorex™, Finnpipette®, Pipetman®, and Rainin® series pipettors.
Certifications: Certified RNase-, DNase-, DNA-, and pyrogen-free.
Packaging: Tips are packaged 96/tray, 8 trays per pack.