Nalgene® Closed-Dome Bio Tank Closure with Mixer Support, Thermo Scientific
Supplier: Thermo Fisher Scientific
2651-0200
66463-210CS 791.41 USD
66463-210
Nalgene® Closed-Dome Bio Tank Closure with Mixer Support, Thermo Scientific
Bioreactors
Closure with overhead mixer support for use with Closed-Dome Bio Tanks (see 66463-206 series)
Sanitary flange allows aseptic mixing in a closed system. The assembly consists of a 185mm polypropylene screw closure with a 2" sanitary fitting ferrule welded in the center, a 2" silicone gasket, and a true union fitting. Can be connected to other 2" sanitary fittings for drain lines and closed system filling. Individually packaged. Autoclavable, but must be kept vertical if assembled with mixer