Rocker II™ Platform Mixer, Boekel Scientific
Supplier: Boekel
260350
66238-336EA 1493.37 USD
66238-336
Rocker II™ Platform Mixer, Boekel Scientific
Shakers and Mixers Rotating/Rocking Shakers
Front to back rocking motion is ideal for gel staining and destaining, Northern, Western, and Southern blots, mixing test tubes, and rocking agglutination cards.
  • Tilt angle is adjustable from 0–20°
  • Speed is adjustable from 3–25 strokes per minute
  • Both tilt angle and speed are adjustable without tools
Rocker accepts a wide variety of flasks, plates, and tubes. Thin power cord easily passes through incubator door gaskets.
Certifications: The 230V model is CE marked.
Ordering information: Supplied with flat, non-skid mat. Low voltage (12V) power is supplied by wall mount transformer (included). Accessories sold separately.